6,7-Difluoro-1,4-dihydroquinoxaline-2,3-dione

Lieferant: Apollo Scientific

Synonyme: 6,7-Difluoroquinoxaline-2,3(1H,4H)-dione

PC210109-1G
APOSPC210109-1GEA 202 EUR
APOSPC210109-1G
6,7-Difluoro-1,4-dihydroquinoxaline-2,3-dione
6,7-Difluoro-1,4-dihydroquinoxaline-2,3-dione
Formel: C₈H₄F₂N₂O₂
Molecular Weight: 198,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00907424
CAS-Nummer: 91895-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR