(Z)-N-{2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1-(3,4-dichlorophenyl)ethylidene}hydroxylamine

Lieferant: Apollo Scientific

PC210112-500MG
APOSPC210112-500MGEA 640 EUR
APOSPC210112-500MG
(Z)-N-{2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1-(3,4-dichlorophenyl)ethylidene}hydroxylamine
(Z)-N-{2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1-(3,4-dichlorophenyl)ethylidene}hydroxylamine
Formel: C₁₄H₈Cl₃F₃N₂O
Molecular Weight: 383,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28962695
CAS-Nummer: 1796581-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR