Ethyl 2-methyl-5-(((trifluoromethyl)-sulphonyl)-oxy)-1H-indole-3-carboxylate

Lieferant: Apollo Scientific

PC210113-500MG
APOSPC210113-500MGEA 430 EUR
APOSPC210113-500MG
Ethyl 2-methyl-5-(((trifluoromethyl)-sulphonyl)-oxy)-1H-indole-3-carboxylate
Ethyl 2-methyl-5-(((trifluoromethyl)-sulphonyl)-oxy)-1H-indole-3-carboxylate
Formel: C₁₃H₁₂F₃NO₅S
Molecular Weight: 351,298 g/mol
Schmelzpunkt: 153…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD28962696
CAS-Nummer: 1796574-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR