N'-(5-Chloropentanoyl)-4-fluorobenzohydrazide

Lieferant: Apollo Scientific

PC210114-500MG
APOSPC210114-500MGEA 430 EUR
APOSPC210114-500MG
N'-(5-Chloropentanoyl)-4-fluorobenzohydrazide
N'-(5-Chloropentanoyl)-4-fluorobenzohydrazide
Formel: C₁₂H₁₄ClFN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28976603
CAS-Nummer: 1823188-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR