N'-(3-chloropropanoyl)-4-fluorobenzohydrazide

Lieferant: Apollo Scientific

PC210115-500MG
APOSPC210115-500MGEA 430 EUR
APOSPC210115-500MG
N'-(3-chloropropanoyl)-4-fluorobenzohydrazide
N'-(3-chloropropanoyl)-4-fluorobenzohydrazide
Formel: C₁₀H₁₀ClFN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28976604
CAS-Nummer: 1823188-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR