N'-(3-chloropropanoyl)-3-fluorobenzohydrazide

Lieferant: Apollo Scientific

PC210117-500MG
APOSPC210117-500MGEA 430 EUR
APOSPC210117-500MG
N'-(3-chloropropanoyl)-3-fluorobenzohydrazide
N'-(3-chloropropanoyl)-3-fluorobenzohydrazide
Formel: C₁₀H₁₀ClFN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28976605
CAS-Nummer: 1823182-50-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR