(R)-2-methyl-N-(3-(trifluoromethyl)benzylidene)propane-2-sulphinamide

Lieferant: Apollo Scientific

PC210124-500MG
APOSPC210124-500MGEA 217 EUR
APOSPC210124-500MG
(R)-2-methyl-N-(3-(trifluoromethyl)benzylidene)propane-2-sulphinamide
(R)-2-methyl-N-(3-(trifluoromethyl)benzylidene)propane-2-sulphinamide
Formel: C₁₂H₁₄F₃NOS
Lagertemperatur: Raumtemperatur
MDL: MFCD18781370
CAS-Nummer: 1547490-91-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR