(S)-2-methyl-N-(3-(trifluoromethyl)benzylidene)propane-2-sulphinamide
Lieferant: Apollo Scientific
PC210125-500MG
APOSPC210125-500MGEA
321
EUR
APOSPC210125-500MG
(S)-2-methyl-N-(3-(trifluoromethyl)benzylidene)propane-2-sulphinamide
(S)-2-methyl-N-(3-(trifluoromethyl)benzylidene)propane-2-sulphinamide
Formel:
C₁₂H₁₄F₃NOS Molecular Weight: 277,306 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18737116 CAS-Nummer: 1453070-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...