(R)-2-methyl-N-(2-(trifluoromethyl)benzylidene)propane-2-sulphinamide

Lieferant: Apollo Scientific

PC210126-500MG
APOSPC210126-500MGEA 217 EUR
APOSPC210126-500MG
(R)-2-methyl-N-(2-(trifluoromethyl)benzylidene)propane-2-sulphinamide
(R)-2-methyl-N-(2-(trifluoromethyl)benzylidene)propane-2-sulphinamide
Formel: C₁₂H₁₄F₃NOS
Molecular Weight: 277,306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18736888
CAS-Nummer: 1415337-87-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR