[1-(2,2,2-Trifluoroethyl)-1H-pyrazol-5-yl]methanol ≥98%

Lieferant: Apollo Scientific

Synonyme: (1-(2,2,2-Trifluoroethyl)-1H-pyrazol-5-yl)methanol

PC210127-100MG PC210127-1G PC210127-250MG
APOSPC210127-100MGEA 353 EUR
APOSPC210127-100MG APOSPC210127-1G APOSPC210127-250MG
[1-(2,2,2-Trifluoroethyl)-1H-pyrazol-5-yl]methanol ≥98%
[1-(2,2,2-Trifluoroethyl)-1H-pyrazol-5-yl]methanol
Formel: C₆H₇F₃N₂O
Molecular Weight: 180.128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15976765
CAS-Nummer: 1296225-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR