4-Fluoro-5-iodo-1H-indazole ≥98%

Lieferant: Apollo Scientific

PC210132-1G
APOSPC210132-1GEA 119 EUR
APOSPC210132-1G
4-Fluoro-5-iodo-1H-indazole ≥98%
4-Fluoro-5-iodo-1H-indazole
Formel: C₇H₄FIN₂
Molecular Weight: 262.023 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11007929
CAS-Nummer: 1082041-87-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR