6-Fluoro-5-iodo-1H-indazole ≥98%

Lieferant: Apollo Scientific

PC210133-1G
APOSPC210133-1GEA 29 EUR
APOSPC210133-1G
6-Fluoro-5-iodo-1H-indazole ≥98%
6-Fluoro-5-iodo-1H-indazole
Formel: C₇H₄FIN₂
Molecular Weight: 262.023 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18381157
CAS-Nummer: 1260384-77-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR