4-Iodo-1-(2,2,2-trifluoroethyl)-1H-pyrazole ≥98%
Lieferant: Apollo Scientific
PC210143-250MG
APOSPC210143-250MGEA
94.4
EUR
APOSPC210143-250MG
4-Iodo-1-(2,2,2-trifluoroethyl)-1H-pyrazole ≥98%
4-Iodo-1-(2,2,2-trifluoroethyl)-1H-pyrazole
Formel:
C₅H₄F₃IN₂ Molecular Weight: 275,998 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20327556 CAS-Nummer: 918483-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...