N-(2-Fluoroethyl)aniline ≥98%

Lieferant: Apollo Scientific

PC210144-100MG PC210144-1G PC210144-250MG
APOSPC210144-100MGEA 168 EUR
APOSPC210144-100MG APOSPC210144-1G APOSPC210144-250MG
N-(2-Fluoroethyl)aniline ≥98%
N-(2-Fluoroethyl)aniline
Formel: C₈H₁₀FN
Molecular Weight: 139.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19217349
CAS-Nummer: 459-40-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR