5,6-Difluoro-2,3-dihydro-1H-isoindol-1-one ≥98%
Lieferant: Apollo Scientific
Synonyme:
5,6-Difluoro-2,3-dihydro-1h-isoindol-1-one
PC210145-100MG
PC210145-1G
PC210145-250MG
APOSPC210145-100MGEA
190
EUR
APOSPC210145-100MG
APOSPC210145-1G
APOSPC210145-250MG
5,6-Difluoro-2,3-dihydro-1H-isoindol-1-one ≥98%
5,6-Difluoro-2,3-dihydro-1H-isoindol-1-one
Formel:
C₈H₅F₂NO |
MDL:
MFCD24617103 CAS-Nummer: 1192040-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...