1-(2-Cyanoethyl)-5-(trifluoromethyl)pyrid-2-(1H)-one
Lieferant: Apollo Scientific
|
Achtung
|
PC2108-1G
PC2108-10G
APOSPC2108-1GEA
29
EUR
APOSPC2108-1G
APOSPC2108-10G
1-(2-Cyanoethyl)-5-(trifluoromethyl)pyrid-2-(1H)-one
1-(2-Cyanoethyl)-5-(trifluoromethyl)pyrid-2-(1H)-one
Formel:
C₉H₇F₃N₂O Schmelzpunkt: 89…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00204218 CAS-Nummer: 175277-71-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...