Cyclopropylmethyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2115-100MG
APOSPC2115-100MGEA
0
EUR
APOSPC2115-100MG
Cyclopropylmethyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate 97%
Cyclopropylmethyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
Formel:
C₁₀H₈ClF₃N₂O₂ Siedepunkt: 108…110 °C (0,5 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068151 CAS-Nummer: 175137-31-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...