6-Fluor-1-indanon 97%
Lieferant: Apollo Scientific
PC2137-100G
APOSPC2137-100GEA
229
EUR
APOSPC2137-100G
6-Fluor-1-indanon 97%
6-Fluor-1-indanon
Formel:
C₉H₇FO Molecular Weight: 150,15 g/mol Siedepunkt: 60 °C/0,5mmHg Schmelzpunkt: 53…55 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318147 CAS-Nummer: 1481-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...