N-Benzyl-2,3-difluoro-4-(trifluoromethyl)aniline 98%

Lieferant: Apollo Scientific

PC2164-1G
APOSPC2164-1GEA 807 EUR
APOSPC2164-1G
N-Benzyl-2,3-difluoro-4-(trifluoromethyl)aniline 98%
N-Benzyl-2,3-difluoro-4-(trifluoromethyl)aniline
Formel: C₁₄H₁₀F₅N
Molecular Weight: 287.228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972668
CAS-Nummer: 887267-93-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR