1,2-Diamino-4-chloro-5-fluorobenzene 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Chloro-5-fluorobenzene-1,2-diamine

PC2182-1G PC2182-25G PC2182-5G
APOSPC2182-1GEA 21.9 EUR
APOSPC2182-1G APOSPC2182-25G APOSPC2182-5G
1,2-Diamino-4-chloro-5-fluorobenzene 98%
1,2-Diamino-4-chloro-5-fluorobenzene
Formel: C₆H₆ClFN₂
Molecular Weight: 160.58 g/mol
Schmelzpunkt: 110…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042485
CAS-Nummer: 139512-70-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR