2,2-Bis(3-aminophenyl)hexafluoropropane 98%
Lieferant: Apollo Scientific
Synonyme:
3,3'-(Hexafluoroisopropylidene)dianiline
PC2186-5G
APOSPC2186-5GEA
690
EUR
APOSPC2186-5G
2,2-Bis(3-aminophenyl)hexafluoropropane 98%
2,2-Bis(3-aminophenyl)hexafluoropropane
Formel:
C₁₅H₁₂F₆N₂ Molecular Weight: 334,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191619 CAS-Nummer: 47250-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...