4-Fluor-o-phenylendiamin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluorbenzol-1,2-diamin, 1,2-Diamino-4-fluorbenzol, 3,4-Diamino-1-fluorbenzol, 4-Fluor-1,2-phenylendiamin

PC2187-25G
APOSPC2187-25GEA 45.1 EUR
APOSPC2187-25G
4-Fluor-o-phenylendiamin 97%
4-Fluor-o-phenylendiamin

Intermediate for the synthesis of poly(arylene ether phenylquinoxanlines): ACS Symp. Ser. 407, 212, 1999

Formel: C₆H₇FN₂
Molecular Weight: 126,13 g/mol
Schmelzpunkt: 94…98 °C
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00042228
CAS-Nummer: 367-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR