2,4-Diamino-6-(4-Fluorophenyl)Pyrimidine 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2188-1G
APOSPC2188-1GEA
34.8
EUR
APOSPC2188-1G
2,4-Diamino-6-(4-Fluorophenyl)Pyrimidine 97%
2,4-Diamino-6-(4-Fluorophenyl)Pyrimidine
Formel:
C₁₀H₉FN₄ Schmelzpunkt: 164…166 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052076 CAS-Nummer: 175137-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...