1-(4-Nitrophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

PC2202-1G
APOSPC2202-1GEA 45.1 EUR
APOSPC2202-1G
1-(4-Nitrophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid 97%
1-(4-Nitrophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid
Formel: C₁₁H₆F₃N₃O₄
Molecular Weight: 301,18 g/mol
Schmelzpunkt: 202…204 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173867
CAS-Nummer: 142818-03-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR