2-Amino-5,6-bis(difluoromethoxy)-1,3-benzothiazole 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5,6-Bis(difluoromethoxy)benzo[d]thiazol-2-amine, 5,6-Bis(difluoromethoxy)-1,3-benzothiazol-2-amine

PC2205-1G
APOSPC2205-1GEA 259 EUR
APOSPC2205-1G
2-Amino-5,6-bis(difluoromethoxy)-1,3-benzothiazole 98%
2-Amino-5,6-bis(difluoromethoxy)-1,3-benzothiazole
Formel: C₉H₆F₄N₂O₂S
Molecular Weight: 282,215 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972671
CAS-Nummer: 887268-01-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR