1,1'-(1,1,2,2-Tetrafluoroethane-1,2-diyl)bis(4-fluorobenzene) 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC2215-5G
APOSPC2215-5GEA
483
EUR
APOSPC2215-5G
1,1'-(1,1,2,2-Tetrafluoroethane-1,2-diyl)bis(4-fluorobenzene) 98%
1,1'-(1,1,2,2-Tetrafluoroethane-1,2-diyl)bis(4-fluorobenzene)
Formel:
C₁₄H₈F₆ Schmelzpunkt: 96…97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631642 CAS-Nummer: 4100-99-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...