3-Brom-4-(trifluormethoxy)benzamid 98+%
Lieferant: Apollo Scientific
|
Achtung
|
PC2253-1G
PC2253-250MG
PC2253-5G
APOSPC2253-1GEA
58
EUR
APOSPC2253-1G
APOSPC2253-250MG
APOSPC2253-5G
3-Brom-4-(trifluormethoxy)benzamid 98+%
3-Brom-4-(trifluormethoxy)benzamid
Formel:
C₈H₅BrF₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458067 CAS-Nummer: 914636-29-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...