3-Brom-4-(trifluormethoxy)benzamid 98+%

Lieferant: Apollo Scientific
Achtung

PC2253-1G PC2253-250MG PC2253-5G
APOSPC2253-1GEA 58 EUR
APOSPC2253-1G APOSPC2253-250MG APOSPC2253-5G
3-Brom-4-(trifluormethoxy)benzamid 98+%
3-Brom-4-(trifluormethoxy)benzamid
Formel: C₈H₅BrF₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08458067
CAS-Nummer: 914636-29-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR