2,6-Dibrom-4-fluoranilin 98%

Lieferant: Apollo Scientific
Achtung

PC2255-100G PC2255-25G PC2255-500G
APOSPC2255-100GEA 42.2 EUR
APOSPC2255-100G APOSPC2255-25G APOSPC2255-500G
2,6-Dibrom-4-fluoranilin 98%
2,6-Dibrom-4-fluoranilin
Formel: C₆H₄Br₂FN
Schmelzpunkt: 63…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041445
CAS-Nummer: 344-18-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR