1,2-Dibrom-1-iod-1,2,2-trifluorethan 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2274-5G
APOSPC2274-5GEA
27.7
EUR
APOSPC2274-5G
1,2-Dibrom-1-iod-1,2,2-trifluorethan 97%
1,2-Dibrom-1-iod-1,2,2-trifluorethan
Formel:
C₂Br₂F₃I Siedepunkt: 65 °C/32mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042521 CAS-Nummer: 216394-01-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...