4,4'-Dibromoctafluorbiphenyl (DBOB) 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4,4-Dibromooctafluorobiphenyl
PC2279-1G
PC2279-25G
APOSPC2279-1GEA
62.5
EUR
APOSPC2279-1G
APOSPC2279-25G
4,4'-Dibromoctafluorbiphenyl (DBOB) 99%
4,4'-Dibromoctafluorbiphenyl (DBOB)
Formel:
C₁₂Br₂F₈ Molecular Weight: 455.94 g/mol Schmelzpunkt: 113…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000310 CAS-Nummer: 10386-84-2 EINECS: 233-847-1 UN: 3152 ADR: 9 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...