4,5-Difluorobenzimidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,5-Difluoro-1H-benzo[d]imidazole

PC2313-250MG
APOSPC2313-250MGEA 43.5 EUR
APOSPC2313-250MG
4,5-Difluorobenzimidazole
4,5-Difluorobenzimidazole
Formel: C₇H₄F₂N₂
Molecular Weight: 154,12 g/mol
Schmelzpunkt: 211…213 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00153122
CAS-Nummer: 236736-21-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR