1-Benzyl-4,4-difluorpiperidin

Lieferant: Apollo Scientific
Achtung

PC2356-100MG
APOSPC2356-100MGEA 14.7 EUR
APOSPC2356-100MG
1-Benzyl-4,4-difluorpiperidin
1-Benzyl-4,4-difluorpiperidin
Formel: C₁₂H₁₅F₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD03840201
CAS-Nummer: 155137-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR