trans-1,1,2,2,3,4-Hexafluorocyclobutane 97%

Lieferant: Apollo Scientific

PC2357-5G PC2357-25G
APOSPC2357-5GEA 491 EUR
APOSPC2357-5G APOSPC2357-25G
trans-1,1,2,2,3,4-Hexafluorocyclobutane 97%
trans-1,1,2,2,3,4-Hexafluorocyclobutane
Formel: C₄H₂F₆
Molecular Weight: 164.05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094154
CAS-Nummer: 23012-94-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR