1,2-Dichlor-3-fluorbenzol 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC2398L-1G
PC2398L-25G
PC2398L-5G
APOSPC2398L-1GEA
21.9
EUR
APOSPC2398L-1G
APOSPC2398L-25G
APOSPC2398L-5G
1,2-Dichlor-3-fluorbenzol 99%
1,2-Dichloro-3-fluorobenzene
Formel:
C₆H₃Cl₂F Siedepunkt: 80 °C/22mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060647 CAS-Nummer: 36556-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...