4,5-Dichloro-2-(2-fluorobenzyl)pyridazine-3(2H)-one 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2404-250MG
APOSPC2404-250MGEA
43.5
EUR
APOSPC2404-250MG
4,5-Dichloro-2-(2-fluorobenzyl)pyridazine-3(2H)-one 97%
4,5-Dichloro-2-(2-fluorobenzyl)pyridazine-3(2H)-one
Formel:
C₁₁H₇Cl₂FN₂O Molecular Weight: 273,09 g/mol Schmelzpunkt: 110…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067775 CAS-Nummer: 175135-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...