1,1-Difluoropropane (FC-272fb) 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC2405-5G
APOSPC2405-5GEA
720
EUR
APOSPC2405-5G
1,1-Difluoropropane (FC-272fb) 97%
1,1-Difluoropropane (FC-272fb)
Formel:
C₃H₆F₂ Molecular Weight: 80,077 g/mol Siedepunkt: 44415 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258968 CAS-Nummer: 430-61-5 UN: 3161 ADR: 2.1, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...