2-Bromo-4-(methoxymethyl)fluorobenzene 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-1-fluoro-4-(methoxymethyl)benzene

PC2422-1G
APOSPC2422-1GEA 104 EUR
APOSPC2422-1G
2-Bromo-4-(methoxymethyl)fluorobenzene 95+%
2-Bromo-4-(methoxymethyl)fluorobenzene
Formel: C₈H₈BrFO
Molecular Weight: 219.051 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04116324
CAS-Nummer: 887268-22-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR