2-Methyl-5-(trifluoromethyl)benzyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-(Chloromethyl)-4-methylbenzotrifluoride, 2-(Chloromethyl)-4-(trifluoromethyl)toluene
PC2436-250MG
APOSPC2436-250MGEA
215
EUR
APOSPC2436-250MG
2-Methyl-5-(trifluoromethyl)benzyl chloride
2-Methyl-5-(trifluoromethyl)benzyl chloride
Formel:
C₉H₈ClF₃ Molecular Weight: 208,608 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861155 CAS-Nummer: 225656-63-3 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...