Ethyl 2-bromo-2-fluoro-3-oxo-3-phenylpropanoate 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC2439-1G
APOSPC2439-1GEA
603
EUR
APOSPC2439-1G
Ethyl 2-bromo-2-fluoro-3-oxo-3-phenylpropanoate 98%
Ethyl 2-bromo-2-fluoro-3-oxo-3-phenylpropanoate
Formel:
C₁₁H₁₀BrFO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03844745 CAS-Nummer: 139101-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...