5-Methyl-2-(trifluoromethoxy)bromobenzene 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-4-methyl-1-(trifluoromethoxy)benzene

PC2455-1G
APOSPC2455-1GEA 186 EUR
APOSPC2455-1G
5-Methyl-2-(trifluoromethoxy)bromobenzene 95+%
5-Methyl-2-(trifluoromethoxy)bromobenzene
Formel: C₈H₆BrF₃O
Molecular Weight: 255,032 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972699
CAS-Nummer: 887268-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR