[4-(Trifluoromethyl)phenylthio]acetic acid 97%

Lieferant: Apollo Scientific
Achtung

PC2456-250MG
APOSPC2456-250MGEA 29 EUR
APOSPC2456-250MG
[4-(Trifluoromethyl)phenylthio]acetic acid 97%
[4-(Trifluoromethyl)phenylthio]acetic acid
Formel: C₉H₇F₃O₂S
Molecular Weight: 236,21 g/mol
Schmelzpunkt: 114…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00236351
CAS-Nummer: 102582-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR