4-Brom-2,3,5,6-tetrafluorbenzonitril 98%

Lieferant: Apollo Scientific
Achtung

PC2466-1G PC2466-10G
APOSPC2466-1GEA 21.9 EUR
APOSPC2466-1G APOSPC2466-10G
4-Brom-2,3,5,6-tetrafluorbenzonitril 98%
4-Brom-2,3,5,6-tetrafluorbenzonitril
Formel: C₇BrF₄N
Molecular Weight: 253.9792 g/mol
Schmelzpunkt: 77…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075282
CAS-Nummer: 17823-40-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR