3-Brom-4-(trifluormethoxy)benzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Bromo-4-(trifluoromethoxy)phenyl)methanol

PC2473-1G PC2473-250MG PC2473-5G
APOSPC2473-1GEA 174 EUR
APOSPC2473-1G APOSPC2473-250MG APOSPC2473-5G
3-Brom-4-(trifluormethoxy)benzylalkohol 98%
3-Brom-4-(trifluormethoxy)benzylalkohol
Formel: C₈H₆BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07777125
CAS-Nummer: 85366-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR