2,4-Dichlor-3-nitrobenzotrifluorid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3-Dichloro-2-nitro-4-(trifluoromethyl)benzene, 2,4-Dichloro-3-nitrobenzotrifluoride

PC2487-5G
APOSPC2487-5GEA 495 EUR
APOSPC2487-5G
2,4-Dichlor-3-nitrobenzotrifluorid
2,4-Dichlor-3-nitrobenzotrifluorid
Formel: C₇H₂Cl₂F₃NO₂
Molecular Weight: 260 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00153114
CAS-Nummer: 203915-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR