2,4-Dichlor-5-nitrobenzotrifluorid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4-Dichloro-5-nitrobenzotrifluoride, 1,3-Dichloro-5-nitro-4-(trifluoromethyl)benzene
PC2488-25G
APOSPC2488-25GEA
23.2
EUR
APOSPC2488-25G
2,4-Dichlor-5-nitrobenzotrifluorid 98%
2,4-Dichlor-5-nitrobenzotrifluorid
Formel:
C₇H₂Cl₂F₃NO₂ Molecular Weight: 260 g/mol Schmelzpunkt: 53…57 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007073 CAS-Nummer: 400-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...