4'-Butyl-2,2,2-trifluoracetophenon (Carbonat Ionophor IV ) 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Butyl-2,2,2-trifluoroacetophenone, 1-(4-Butylphenyl)-2,2,2-trifluoroethanone

PC2499-1G
APOSPC2499-1GEA 408 EUR
APOSPC2499-1G
4'-Butyl-2,2,2-trifluoracetophenon (Carbonat Ionophor IV ) 97%
4'-Butyl-2,2,2-trifluoracetophenon (Carbonat Ionophor IV )
Formel: C₁₂H₁₃F₃O
Molecular Weight: 230.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00214157
CAS-Nummer: 40739-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR