Fluoromethyl 4-methylbenzenesulphonate 97+%

Lieferant: Apollo Scientific
Gefahr

PC250002-1G
APOSPC250002-1GEA 74.1 EUR
APOSPC250002-1G
Fluoromethyl 4-methylbenzenesulphonate 97+%
Fluoromethyl 4-methylbenzenesulphonate
Formel: C₈H₉FO₃S
Molecular Weight: 204.219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20482732
CAS-Nummer: 114435-86-8
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR