4-Fluoroisoquinoline 95+%

Lieferant: Apollo Scientific

PC250003-10G PC250003-1G PC250003-5G
APOSPC250003-10GEA 440 EUR
APOSPC250003-10G APOSPC250003-1G APOSPC250003-5G
4-Fluoroisoquinoline 95+%
4-Fluoroisoquinoline
Formel: C₉H₆FN
Molecular Weight: 147.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06656128
CAS-Nummer: 394-67-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR