2-(Trifluoromethyl)-9H-carbazole 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC250007-100MG
APOSPC250007-100MGEA
509
EUR
APOSPC250007-100MG
2-(Trifluoromethyl)-9H-carbazole 98%
2-(Trifluoromethyl)-9H-carbazole
Formel:
C₁₃H₈F₃N Molecular Weight: 235,205 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09033509 CAS-Nummer: 2285-35-0 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...